The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145128.2 | NP_001138600.2 | 6 - 7 | 523 | 123 |
NM_145025.4 | NP_659462.1 | 6 - 7 | 523 | 123 | |
XM_006715376.3 | XP_006715439.2 | 7 - 8 | 1388 | 123 | |
XM_011535550.2 | XP_011533852.1 | 7 - 8 | 1388 | 123 | |
XM_011535552.2 | XP_011533854.1 | 6 - 7 | 564 | 123 | |
XM_011535553.2 | XP_011533855.1 | 6 - 7 | 1171 | 123 | |
XM_011535554.2 | XP_011533856.1 | 7 - 8 | 1388 | 123 | |
XM_011535555.2 | XP_011533857.1 | 7 - 8 | 1388 | 123 | |
XM_011535556.2 | XP_011533858.1 | 7 - 8 | 1388 | 123 | |
XM_011535559.2 | XP_011533861.1 | 7 - 8 | 1388 | 123 | |
XM_017010384.1 | XP_016865873.1 | 7 - 8 | 1388 | 123 | |
XM_017010385.1 | XP_016865874.1 | 6 - 7 | 541 | 123 | |
XM_017010386.1 | XP_016865875.1 | 7 - 8 | 1388 | 123 | |
XM_017010387.1 | XP_016865876.1 | 1 - 2 | 109 | 123 | |
XM_017010388.1 | XP_016865877.1 | 7 - 8 | 1388 | 123 | |
GenBank mRNA | AK055538.1 | - | 6 - 7 | 500 | 123 |
AK315561.1 | - | 6 - 7 | 494 | 123 | |
AL832162.1 | - | 6 - 7 | 502 | 123 | |
BC022031.2 | - | 6 - 7 | 528 | 123 |