The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017539.2 | NP_060009.1 | 7 - 8 | 1080 | 79 |
XM_006721062.1 | XP_006721125.1 | 8 - 9 | 1269 | 79 | |
XM_011545882.2 | XP_011544184.1 | 9 - 10 | 1709 | 79 | |
XM_011545883.1 | XP_011544185.1 | 9 - 10 | 1477 | 79 | |
XM_011545885.2 | XP_011544187.1 | 9 - 10 | 1709 | 79 | |
XM_017023426.1 | XP_016878915.1 | 9 - 10 | 1709 | 79 | |
XM_017023427.1 | XP_016878916.1 | 9 - 10 | 1709 | 79 | |
XM_017023428.1 | XP_016878917.1 | 9 - 10 | 1709 | 79 | |
XM_017023429.1 | XP_016878918.1 | 9 - 10 | 1710 | 79 | |
XM_017023431.1 | XP_016878920.1 | 9 - 10 | 1710 | 79 | |
XM_017023432.1 | XP_016878921.1 | 7 - 8 | 1176 | 79 | |
GenBank mRNA | AF494040.1 | - | 7 - 8 | 1080 | 79 |
AK303958.1 | - | 8 - 9 | 1074 | 79 |