The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002263.3 | NP_002254.2 | 7 - 8 | 1991 | 59 |
XM_011514585.1 | XP_011512887.1 | 7 - 8 | 1697 | 59 | |
XM_017010836.1 | XP_016866325.1 | 7 - 8 | 1697 | 59 | |
XM_017010837.1 | XP_016866326.1 | 7 - 8 | 1806 | 59 | |
GenBank mRNA | AK297378.1 | - | 7 - 8 | 1604 | 59 |
AK303239.1 | - | 7 - 8 | 1631 | 59 | |
BC000712.2 | - | 7 - 8 | 1699 | 59 | |
BC063567.1 | - | 7 - 8 | 1649 | 59 | |
BC073878.1 | - | 7 - 8 | 1684 | 59 | |
BC098438.1 | - | 7 - 8 | 1681 | 59 | |
BC121041.1 | - | 7 - 8 | 1622 | 59 | |
BC121042.1 | - | 7 - 8 | 1622 | 59 | |
D14678.1 | - | 2 - 3 | 1081 | 59 |