The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012241.1 | NP_001012241.1 | 1 - 2 | 224 | 62 |
GenBank mRNA | BC118997.1 | - | 1 - 2 | 147 | 62 |
BC122543.1 | - | 1 - 2 | 147 | 62 |