The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021089.2 | NP_066575.2 | 3 - 4 | 421 | 74 |
GenBank mRNA | AK093110.1 | - | 3 - 4 | 421 | 74 |
AK225825.1 | - | 3 - 4 | 418 | 74 | |
BC039323.1 | - | 3 - 4 | 379 | 74 | |
M29581.1 | - | 2 - 3 | 195 | 74 |