Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KPNA3
Entrez Gene ID
3839
Gene Name
karyopherin subunit alpha 3
Gene Aliases
IPOA4, SRP1, SRP1gamma, SRP4, hSRP1
Chromosome Location
Chr.13: 49699307 - 49792921 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002267.3 NP_002258.2 9 - 10 1138 129
XM_017020561.1 XP_016876050.1 9 - 10 821 129
GenBank mRNA AF005263.1 - 9 - 10 735 129
AK223480.1 - 9 - 10 760 129
AK290528.1 - 9 - 10 798 129
AK291000.1 - 9 - 10 762 129
BC017355.2 - 9 - 10 747 129
BC024202.2 - 9 - 10 747 129
BC035090.2 - 9 - 10 1139 129
D89618.1 - 9 - 10 814 129

View Full Product Details