The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001764.2 | NP_001755.1 | 4 - 5 | 995 | 91 |
GenBank mRNA | AK303330.1 | - | 4 - 5 | 902 | 91 |
BC069481.1 | - | 4 - 5 | 932 | 91 | |
BC074747.2 | - | 4 - 5 | 909 | 91 | |
BC104216.1 | - | 4 - 5 | 932 | 91 | |
BC104217.1 | - | 4 - 5 | 932 | 91 | |
M28826.1 | - | 4 - 5 | 932 | 91 |