The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004782.3 | NP_004773.1 | 2 - 3 | 542 | 97 |
GenBank mRNA | AF115436.1 | - | 2 - 3 | 509 | 97 |
BC009715.1 | - | 2 - 3 | 506 | 97 | |
BG773022.1 | - | 2 - 3 | 545 | 97 | |
BT007357.1 | - | 2 - 3 | 435 | 97 | |
CR456582.1 | - | 2 - 3 | 468 | 97 |