Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AK9
Entrez Gene ID
221264
Gene Name
adenylate kinase 9
Gene Aliases
AK 9, AKD1, AKD2, C6orf199, C6orf224, dJ70A9.1
Chromosome Location
Chr.6: 109492855 - 109691253 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145128.2 NP_001138600.2 15 - 16 1691 163
XM_006715376.3 XP_006715439.2 16 - 17 2556 163
XM_011535550.2 XP_011533852.1 16 - 17 2556 163
XM_011535552.2 XP_011533854.1 15 - 16 1732 163
XM_011535553.2 XP_011533855.1 15 - 16 2339 163
XM_011535554.2 XP_011533856.1 14 - 15 2235 163
XM_011535555.2 XP_011533857.1 16 - 17 2556 163
XM_011535556.2 XP_011533858.1 16 - 17 2556 163
XM_011535559.2 XP_011533861.1 16 - 17 2556 163
XM_017010384.1 XP_016865873.1 16 - 17 2556 163
XM_017010385.1 XP_016865874.1 15 - 16 1724 163
XM_017010386.1 XP_016865875.1 16 - 17 2556 163
XM_017010387.1 XP_016865876.1 10 - 11 2187 163
XM_017010388.1 XP_016865877.1 16 - 17 2556 163
GenBank mRNA AK124171.1 - 3 - 4 325 163

View Full Product Details