The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301030.1 | NP_001287959.1 | 13 - 14 | 1739 | 67 |
NM_002088.4 | NP_002079.3 | 13 - 14 | 1729 | 67 | |
XM_005258821.3 | XP_005258878.1 | 14 - 15 | 2642 | 67 | |
XM_011526862.2 | XP_011525164.1 | 14 - 15 | 2645 | 67 | |
XM_011526863.2 | XP_011525165.1 | 14 - 15 | 2642 | 67 | |
XM_011526865.1 | XP_011525167.1 | 15 - 16 | 3146 | 67 | |
XM_011526867.1 | XP_011525169.1 | 16 - 17 | 3368 | 67 | |
XM_011526868.1 | XP_011525170.1 | 15 - 16 | 3124 | 67 | |
XM_011526869.2 | XP_011525171.1 | 14 - 15 | 2645 | 67 | |
XM_017026713.1 | XP_016882202.1 | 16 - 17 | 3371 | 67 | |
GenBank mRNA | AJ249209.1 | - | 13 - 14 | 1729 | 67 |
BC037358.1 | - | 12 - 13 | 1482 | 67 | |
BC052975.1 | - | 3 - 4 | 571 | 67 | |
S40369.1 | - | 13 - 14 | 1694 | 67 |