The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301030.1 | NP_001287959.1 | 16 - 17 | 2306 | 67 |
NM_002088.4 | NP_002079.3 | 16 - 17 | 2296 | 67 | |
XM_005258821.3 | XP_005258878.1 | 17 - 18 | 3209 | 67 | |
XM_011526862.2 | XP_011525164.1 | 17 - 18 | 3212 | 67 | |
XM_011526863.2 | XP_011525165.1 | 17 - 18 | 3209 | 67 | |
XM_011526865.1 | XP_011525167.1 | 18 - 19 | 3713 | 67 | |
XM_011526867.1 | XP_011525169.1 | 19 - 20 | 3935 | 67 | |
XM_011526868.1 | XP_011525170.1 | 18 - 19 | 3691 | 67 | |
XM_017026713.1 | XP_016882202.1 | 19 - 20 | 3938 | 67 | |
GenBank mRNA | AJ249209.1 | - | 16 - 17 | 2296 | 67 |
BC037358.1 | - | 15 - 16 | 2049 | 67 | |
BC052975.1 | - | 6 - 7 | 1138 | 67 | |
S40369.1 | - | 16 - 17 | 2261 | 67 |