The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005258821.3 | XP_005258878.1 | 1 - 2 | 900 | 80 |
XM_011526862.2 | XP_011525164.1 | 1 - 2 | 900 | 80 | |
XM_011526863.2 | XP_011525165.1 | 1 - 2 | 900 | 80 | |
XM_011526865.1 | XP_011525167.1 | 2 - 3 | 1401 | 80 | |
XM_011526867.1 | XP_011525169.1 | 2 - 3 | 1401 | 80 | |
XM_011526868.1 | XP_011525170.1 | 2 - 3 | 1401 | 80 | |
XM_011526869.2 | XP_011525171.1 | 1 - 2 | 900 | 80 | |
XM_011526870.2 | XP_011525172.1 | 1 - 2 | 900 | 80 | |
XM_011526871.2 | XP_011525173.1 | 1 - 2 | 900 | 80 | |
XM_017026713.1 | XP_016882202.1 | 2 - 3 | 1401 | 80 |