Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GRIK5
Entrez Gene ID
2901
Gene Name
glutamate ionotropic receptor kainate type subunit 5
Gene Aliases
EAA2, GRIK2, GluK5, KA2
Chromosome Location
Chr.19: 41998316 - 42071804 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301030.1 NP_001287959.1 1 - 2 126 60
NM_002088.4 NP_002079.3 1 - 2 116 60
XM_005258821.3 XP_005258878.1 2 - 3 1029 60
XM_011526862.2 XP_011525164.1 2 - 3 1029 60
XM_011526863.2 XP_011525165.1 2 - 3 1029 60
XM_011526865.1 XP_011525167.1 3 - 4 1530 60
XM_011526867.1 XP_011525169.1 3 - 4 1530 60
XM_011526869.2 XP_011525171.1 2 - 3 1029 60
XM_011526870.2 XP_011525172.1 2 - 3 1029 60
XM_011526871.2 XP_011525173.1 2 - 3 1029 60
XM_017026713.1 XP_016882202.1 3 - 4 1530 60
GenBank mRNA AJ249209.1 - 1 - 2 116 60
S40369.1 - 1 - 2 81 60

View Full Product Details