The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301030.1 | NP_001287959.1 | 6 - 7 | 787 | 66 |
NM_002088.4 | NP_002079.3 | 6 - 7 | 777 | 66 | |
XM_005258821.3 | XP_005258878.1 | 7 - 8 | 1690 | 66 | |
XM_011526862.2 | XP_011525164.1 | 7 - 8 | 1690 | 66 | |
XM_011526863.2 | XP_011525165.1 | 7 - 8 | 1690 | 66 | |
XM_011526865.1 | XP_011525167.1 | 8 - 9 | 2191 | 66 | |
XM_011526867.1 | XP_011525169.1 | 9 - 10 | 2416 | 66 | |
XM_011526868.1 | XP_011525170.1 | 8 - 9 | 2169 | 66 | |
XM_011526869.2 | XP_011525171.1 | 7 - 8 | 1690 | 66 | |
XM_011526870.2 | XP_011525172.1 | 7 - 8 | 1690 | 66 | |
XM_011526871.2 | XP_011525173.1 | 7 - 8 | 1690 | 66 | |
XM_017026713.1 | XP_016882202.1 | 9 - 10 | 2416 | 66 | |
GenBank mRNA | AJ249209.1 | - | 6 - 7 | 777 | 66 |
BC037358.1 | - | 5 - 6 | 530 | 66 | |
S40369.1 | - | 6 - 7 | 742 | 66 |