The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163940.1 | NP_001157412.1 | 14 - 15 | 1847 | 105 |
NM_002863.4 | NP_002854.3 | 15 - 16 | 1949 | 105 | |
GenBank mRNA | AF046785.1 | - | 15 - 16 | 1822 | 105 |
AF066858.1 | - | 15 - 16 | 1822 | 105 | |
AK225904.1 | - | 15 - 16 | 1902 | 105 | |
AK300580.1 | - | 14 - 15 | 1800 | 105 | |
AK309333.1 | - | 15 - 16 | 1875 | 105 | |
AK313206.1 | - | 15 - 16 | 1863 | 105 | |
BC009895.2 | - | 15 - 16 | 1892 | 105 | |
BC082229.1 | - | 15 - 16 | 1893 | 105 | |
BC095850.1 | - | 15 - 16 | 1909 | 105 | |
BC110791.1 | - | 15 - 16 | 1891 | 105 | |
M14636.1 | - | 15 - 16 | 1935 | 105 | |
M36807.1 | - | 4 - 5 | 379 | 105 | |
Y15233.1 | - | 15 - 16 | 1873 | 105 |