Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PYGL
Entrez Gene ID
5836
Gene Name
phosphorylase, glycogen, liver
Gene Aliases
GSD6
Chromosome Location
Chr.14: 50905217 - 50944530 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002863.4 NP_002854.3 1 - 2 375 67
GenBank mRNA AF046785.1 - 1 - 2 248 67
AK225904.1 - 1 - 2 328 67
AK309333.1 - 1 - 2 301 67
AK313206.1 - 1 - 2 289 67
AL134906.1 - 1 - 2 361 67
BC009895.2 - 1 - 2 318 67
BC065011.1 - 1 - 2 315 67
BC082229.1 - 1 - 2 319 67
BC095850.1 - 1 - 2 335 67
BC110791.1 - 1 - 2 317 67
Y15233.1 - 1 - 2 299 67

View Full Product Details