The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024693.4 | NP_078969.2 | 3 - 4 | 616 | 89 |
GenBank mRNA | AF275677.1 | - | 3 - 4 | 590 | 89 |
AK024562.1 | - | 3 - 4 | 529 | 89 | |
AK222511.1 | - | 3 - 4 | 555 | 89 | |
AK290902.1 | - | 3 - 4 | 546 | 89 | |
BC001091.1 | - | 3 - 4 | 349 | 89 | |
BC009617.1 | - | 3 - 4 | 538 | 89 |