The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270452.1 | NP_001257381.1 | 1 - 2 | 174 | 64 |
NM_006799.3 | NP_006790.1 | 1 - 2 | 174 | 64 | |
NM_144956.2 | NP_659205.1 | 1 - 2 | 174 | 64 | |
NM_144957.2 | NP_659206.1 | 1 - 2 | 174 | 64 | |
NR_073012.1 | 1 - 2 | 174 | 64 | ||
XM_017022875.1 | XP_016878364.1 | 1 - 2 | 98 | 64 | |
XM_017022876.1 | XP_016878365.1 | 1 - 2 | 94 | 64 | |
XM_017022877.1 | XP_016878366.1 | 1 - 2 | 83 | 64 | |
XM_017022878.1 | XP_016878367.1 | 1 - 2 | 94 | 64 | |
GenBank mRNA | AB031329.1 | - | 1 - 2 | 77 | 64 |
AB031330.1 | - | 1 - 2 | 71 | 64 | |
AB031331.1 | - | 1 - 2 | 157 | 64 | |
AF058300.1 | - | 1 - 2 | 86 | 64 | |
AY359034.1 | - | 1 - 2 | 87 | 64 | |
BC074999.2 | - | 1 - 2 | 110 | 64 | |
BC075000.2 | - | 1 - 2 | 110 | 64 | |
BG717268.1 | - | 1 - 2 | 61 | 64 | |
BI827608.1 | - | 1 - 2 | 46 | 64 |