The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032289.2 | NP_115665.1 | 1 - 2 | 156 | 117 |
GenBank mRNA | AL136559.1 | - | 1 - 2 | 156 | 117 |
BC038233.1 | - | 1 - 2 | 133 | 117 |