The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032289.2 | NP_115665.1 | 6 - 7 | 1409 | 77 |
XM_011537696.2 | XP_011535998.1 | 7 - 8 | 1465 | 77 | |
XM_017009975.1 | XP_016865464.1 | 7 - 8 | 2772 | 77 | |
XM_017009976.1 | XP_016865465.1 | 7 - 8 | 3580 | 77 | |
XM_017009977.1 | XP_016865466.1 | 7 - 8 | 3460 | 77 | |
GenBank mRNA | AL136559.1 | - | 6 - 7 | 1409 | 77 |
AL834268.1 | - | 2 - 3 | 191 | 77 | |
BC038233.1 | - | 6 - 7 | 1386 | 77 |