Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARSF
Entrez Gene ID
416
Gene Name
arylsulfatase F
Gene Aliases
ASF
Chromosome Location
Chr.X: 3040234 - 3112729 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201538.1 NP_001188467.1 1 - 2 109 68
GenBank mRNA BP274206.1 - 1 - 2 109 68
BP277542.1 - 1 - 2 107 68
DA628671.1 - 1 - 2 108 68

View Full Product Details