Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AP1G2
Entrez Gene ID
8906
Gene Name
adaptor related protein complex 1 gamma 2 subunit
Gene Aliases
G2AD
Chromosome Location
Chr.14: 23559563 - 23569056 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282474.1 NP_001269403.1 14 - 15 1843 67
NM_001282475.1 NP_001269404.1 14 - 15 1631 67
NM_003917.4 NP_003908.1 16 - 17 1735 67
XM_005268167.3 XP_005268224.1 15 - 16 1858 67
XM_005268168.4 XP_005268225.1 15 - 16 1900 67
XM_005268169.3 XP_005268226.1 15 - 16 1857 67
XM_005268170.3 XP_005268227.1 15 - 16 1857 67
XM_005268172.3 XP_005268229.1 16 - 17 1686 67
XM_005268173.3 XP_005268230.1 16 - 17 1686 67
XM_005268174.3 XP_005268231.1 14 - 15 1733 67
XM_005268175.3 XP_005268232.1 14 - 15 1747 67
XM_005268177.3 XP_005268234.1 15 - 16 1713 67
XM_005268178.3 XP_005268235.1 15 - 16 1604 67
XM_005268179.3 XP_005268236.1 15 - 16 1709 67
XM_005268180.4 XP_005268237.1 14 - 15 1951 67
XM_005268182.3 XP_005268239.1 15 - 16 1580 67
XM_006720301.3 XP_006720364.1 15 - 16 1955 67
XM_011537286.2 XP_011535588.1 7 - 8 979 67
XM_017021740.1 XP_016877229.1 14 - 15 1804 67
XM_017021741.1 XP_016877230.1 12 - 13 1708 67
XM_017021742.1 XP_016877231.1 14 - 15 1775 67
XM_017021746.1 XP_016877235.1 15 - 16 1642 67
XM_017021747.1 XP_016877236.1 15 - 16 3034 67
XM_017021748.1 XP_016877237.1 13 - 14 1662 67
GenBank mRNA AB015318.1 - 16 - 17 1677 67
AF068706.1 - 15 - 16 2398 67
AK226158.1 - 6 - 7 665 67
AK304682.1 - 14 - 15 1466 67
BC051833.1 - 13 - 14 1572 67
BX161425.1 - 14 - 15 1776 67

View Full Product Details