The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282474.1 | NP_001269403.1 | 1 - 2 | 94 | 52 |
NM_003917.4 | NP_003908.1 | 1 - 2 | 94 | 52 | |
XM_005268167.3 | XP_005268224.1 | 1 - 2 | 46 | 52 | |
XM_005268174.3 | XP_005268231.1 | 1 - 2 | 46 | 52 | |
XM_005268177.3 | XP_005268234.1 | 1 - 2 | 197 | 52 | |
XM_017021744.1 | XP_016877233.1 | 1 - 2 | 46 | 52 | |
XM_017021745.1 | XP_016877234.1 | 1 - 2 | 46 | 52 | |
GenBank mRNA | AB015318.1 | - | 1 - 2 | 36 | 52 |
AK307696.1 | - | 1 - 2 | 37 | 52 | |
AK309103.1 | - | 1 - 2 | 46 | 52 | |
BG701521.1 | - | 1 - 2 | 97 | 52 | |
BX161425.1 | - | 1 - 2 | 27 | 52 |