The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020682.3 | NP_065733.2 | 1 - 2 | 145 | 73 |
GenBank mRNA | AF226730.1 | - | 1 - 2 | 82 | 73 |
AK057833.1 | - | 1 - 2 | 164 | 73 | |
AK225972.1 | - | 1 - 2 | 164 | 73 | |
AK309737.1 | - | 1 - 2 | 77 | 73 | |
BC119637.1 | - | 1 - 2 | 49 | 73 | |
BC119638.2 | - | 1 - 2 | 49 | 73 | |
DA368970.1 | - | 1 - 2 | 145 | 73 |