The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319053.1 | NP_001305982.1 | 19 - 20 | 2531 | 62 |
NM_001470.3 | NP_001461.1 | 19 - 20 | 2595 | 62 | |
NM_021903.2 | NP_068703.1 | 14 - 15 | 2395 | 62 | |
NM_021904.3 | NP_068704.2 | 18 - 19 | 2409 | 62 | |
XM_005248982.2 | XP_005249039.1 | 19 - 20 | 2410 | 62 | |
XM_006715047.3 | XP_006715110.1 | 18 - 19 | 2210 | 62 | |
XM_011514453.2 | XP_011512755.1 | 17 - 18 | 2189 | 62 | |
XM_017010676.1 | XP_016866165.1 | 15 - 16 | 1941 | 62 | |
GenBank mRNA | AF099148.1 | - | 19 - 20 | 2417 | 62 |
AJ012185.1 | - | 18 - 19 | 2309 | 62 | |
AJ012186.1 | - | 14 - 15 | 1958 | 62 | |
AJ012187.1 | - | 17 - 18 | 2123 | 62 | |
AJ012288.1 | - | 19 - 20 | 2352 | 62 | |
AJ225028.1 | - | 19 - 20 | 2543 | 62 | |
AJ225029.1 | - | 14 - 15 | 2137 | 62 | |
AK223619.1 | - | 14 - 15 | 2180 | 62 | |
AK290191.1 | - | 14 - 15 | 2180 | 62 | |
AK312308.1 | - | 14 - 15 | 2176 | 62 | |
BC041332.2 | - | 14 - 15 | 2180 | 62 | |
BC042598.1 | - | 14 - 15 | 2169 | 62 | |
BC050532.1 | - | 14 - 15 | 2167 | 62 | |
Y11044.1 | - | 18 - 19 | 2306 | 62 |