The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030641.3 | NP_085144.1 | 1 - 2 | 243 | 63 |
GenBank mRNA | AK095881.1 | - | 1 - 2 | 205 | 63 |
AL834420.1 | - | 1 - 2 | 207 | 63 | |
AX775805.1 | - | 1 - 2 | 215 | 63 | |
AY358210.1 | - | 1 - 2 | 140 | 63 | |
BC038950.2 | - | 1 - 2 | 173 | 63 | |
BC047864.1 | - | 1 - 2 | 225 | 63 | |
BU585026.1 | - | 1 - 2 | 253 | 63 |