The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032588.3 | NP_115977.2 | 3 - 4 | 634 | 67 |
XM_017002559.1 | XP_016858048.1 | 3 - 4 | 634 | 67 | |
GenBank mRNA | AF353673.1 | - | 3 - 4 | 613 | 67 |
AJ276484.1 | - | 3 - 4 | 617 | 67 | |
AK056942.1 | - | 3 - 4 | 630 | 67 | |
AK297820.1 | - | 3 - 4 | 546 | 67 | |
BC080529.1 | - | 3 - 4 | 612 | 67 | |
BT007373.1 | - | 3 - 4 | 494 | 67 |