Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL14A1
Entrez Gene ID
7373
Gene Name
collagen type XIV alpha 1 chain
Gene Aliases
UND
Chromosome Location
Chr.8: 120124468 - 120373575 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021110.3 NP_066933.1 24 - 25 3286 70
XM_005251059.3 XP_005251116.1 24 - 25 3286 70
XM_006716651.3 XP_006716714.1 24 - 25 3149 70
XM_017013809.1 XP_016869298.1 24 - 25 3075 70
GenBank mRNA AK293080.1 - 24 - 25 3256 70
AK307716.1 - 24 - 25 3250 70
AK311498.1 - 24 - 25 3250 70
BC014640.1 - 3 - 4 284 70
BC140893.1 - 23 - 24 3055 70
M64108.1 - 19 - 20 2455 70

View Full Product Details