Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NDE1
Entrez Gene ID
54820
Gene Name
nudE neurodevelopment protein 1
Gene Aliases
HOM-TES-87, LIS4, MHAC, NDE, NUDE, NUDE1
Chromosome Location
Chr.16: 15643267 - 15726353 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143979.1 NP_001137451.1 4 - 5 1062 65
NM_017668.2 NP_060138.1 3 - 4 348 65
XM_005255396.4 XP_005255453.1 3 - 4 341 65
XM_006720897.3 XP_006720960.1 3 - 4 908 65
XM_006720900.3 XP_006720963.1 3 - 4 377 65
XM_011522553.2 XP_011520855.1 4 - 5 1062 65
XM_017023349.1 XP_016878838.1 3 - 4 341 65
XM_017023351.1 XP_016878840.1 3 - 4 341 65
XM_017023352.1 XP_016878841.1 3 - 4 341 65
XM_017023353.1 XP_016878842.1 3 - 4 341 65
XM_017023354.1 XP_016878843.1 3 - 4 341 65
XM_017023355.1 XP_016878844.1 3 - 4 341 65
XM_017023356.1 XP_016878845.1 3 - 4 377 65
GenBank mRNA AF124431.1 - 2 - 3 257 65
AK000108.1 - 3 - 4 327 65
AK025815.1 - 3 - 4 327 65
BC001421.2 - 3 - 4 341 65
BC033900.1 - 4 - 5 1067 65
BP271845.1 - 3 - 4 348 65
KJ534887.1 - 2 - 3 236 65
KJ534967.1 - 2 - 3 236 65
KJ534991.1 - 2 - 3 236 65

View Full Product Details