Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBCD
Entrez Gene ID
6904
Gene Name
tubulin folding cofactor D
Gene Aliases
SSD-1, tfcD
Chromosome Location
Chr.17: 82752048 - 82943186 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005993.4 NP_005984.3 19 - 20 1936 69
XM_005256396.4 XP_005256453.1 18 - 19 1901 69
XM_005256399.4 XP_005256456.1 7 - 8 733 69
XM_005256404.4 XP_005256461.1 7 - 8 775 69
XM_006722290.2 XP_006722353.1 18 - 19 1833 69
XM_006722291.3 XP_006722354.1 7 - 8 1057 69
XM_006722292.2 XP_006722355.1 5 - 6 469 69
XM_011523589.1 XP_011521891.1 14 - 15 1476 69
XM_011523591.1 XP_011521893.1 14 - 15 1461 69
XM_011523593.1 XP_011521895.1 13 - 14 1068 69
XM_011523594.1 XP_011521896.1 6 - 7 538 69
XM_011523595.2 XP_011521897.1 6 - 7 616 69
XM_011523597.2 XP_011521899.1 4 - 5 1269 69
XM_011523598.1 XP_011521900.1 4 - 5 327 69
XM_011523599.2 XP_011521901.1 4 - 5 1045 69
XM_011523600.2 XP_011521902.1 5 - 6 489 69
XM_017024987.1 XP_016880476.1 19 - 20 1797 69
XM_017024988.1 XP_016880477.1 19 - 20 1914 69
XM_017024989.1 XP_016880478.1 3 - 4 183 69
XM_017024990.1 XP_016880479.1 7 - 8 555 69
GenBank mRNA AB023205.1 - 19 - 20 1915 69
AF052173.1 - 5 - 6 324 69
AJ006417.1 - 19 - 20 1848 69
AK057975.1 - 4 - 5 443 69
AK091959.1 - 18 - 19 1822 69
AK293471.1 - 19 - 20 1914 69
AL096745.1 - 3 - 4 323 69
AL133562.1 - 15 - 16 1381 69
BC003094.1 - 19 - 20 1936 69
BC006364.1 - 19 - 20 1893 69
BC039654.1 - 5 - 6 289 69
BC150142.1 - 3 - 4 255 69

View Full Product Details