The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005993.4 | NP_005984.3 | 8 - 9 | 953 | 92 |
XM_005256396.4 | XP_005256453.1 | 7 - 8 | 918 | 92 | |
XM_006722290.2 | XP_006722353.1 | 8 - 9 | 931 | 92 | |
XM_011523590.1 | XP_011521892.1 | 8 - 9 | 931 | 92 | |
XM_017024987.1 | XP_016880476.1 | 8 - 9 | 814 | 92 | |
XM_017024988.1 | XP_016880477.1 | 8 - 9 | 931 | 92 | |
GenBank mRNA | AB023205.1 | - | 8 - 9 | 932 | 92 |
AJ006417.1 | - | 8 - 9 | 865 | 92 | |
AK091959.1 | - | 7 - 8 | 839 | 92 | |
AK293471.1 | - | 8 - 9 | 931 | 92 | |
AL133562.1 | - | 4 - 5 | 398 | 92 | |
BC003094.1 | - | 8 - 9 | 953 | 92 | |
BC006364.1 | - | 8 - 9 | 910 | 92 |