The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002841.1 | NP_001002841.1 | 1 - 2 | 88 | 72 |
XM_011524839.2 | XP_011523141.2 | 1 - 2 | 82 | 72 | |
GenBank mRNA | BU658678.1 | - | 1 - 2 | 94 | 72 |
H83803.1 | - | 1 - 2 | 88 | 72 | |
X52005.1 | - | - | 42 | 72 |