Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP11B
Entrez Gene ID
23200
Gene Name
ATPase phospholipid transporting 11B (putative)
Gene Aliases
ATPIF, ATPIR
Chromosome Location
Chr.3: 182793503 - 182923010 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014616.2 NP_055431.1 29 - 30 3709 79
XM_005247241.2 XP_005247298.1 30 - 31 3794 79
XM_005247243.2 XP_005247300.1 28 - 29 3380 79
XM_011512597.2 XP_011510899.1 27 - 28 3275 79
XM_017006008.1 XP_016861497.1 27 - 28 3272 79
GenBank mRNA AB023173.1 - 16 - 17 1939 79
AK304855.1 - 19 - 20 2602 79
AL133061.1 - 10 - 11 1193 79
BC010630.1 - 15 - 16 1838 79
BX537779.1 - 12 - 13 1325 79

View Full Product Details