The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014616.2 | NP_055431.1 | 8 - 9 | 963 | 76 |
XM_005247241.2 | XP_005247298.1 | 8 - 9 | 962 | 76 | |
XM_011512593.2 | XP_011510895.1 | 8 - 9 | 962 | 76 | |
XM_011512594.2 | XP_011510896.1 | 8 - 9 | 962 | 76 | |
XM_011512597.2 | XP_011510899.1 | 6 - 7 | 529 | 76 | |
XM_017006008.1 | XP_016861497.1 | 6 - 7 | 531 | 76 | |
GenBank mRNA | AF156548.1 | - | 8 - 9 | 872 | 76 |
BX489704.1 | - | 4 - 5 | 378 | 76 |