The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017403.1 | NP_001017403.1 | 4 - 5 | 432 | 95 |
NM_021636.2 | NP_067649.2 | 4 - 5 | 319 | 95 | |
XM_005245404.3 | XP_005245461.1 | 4 - 5 | 376 | 95 | |
XM_011509838.2 | XP_011508140.1 | 4 - 5 | 377 | 95 | |
XM_011509839.2 | XP_011508141.1 | 4 - 5 | 378 | 95 | |
XM_017001996.1 | XP_016857485.1 | 4 - 5 | 378 | 95 | |
GenBank mRNA | AB049405.1 | - | 4 - 5 | 317 | 95 |
AY358119.1 | - | 4 - 5 | 360 | 95 | |
BC047905.1 | - | 4 - 5 | 302 | 95 |