The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144002.1 | NP_001137474.1 | 9 - 10 | 1741 | 81 |
NM_019857.4 | NP_062831.3 | 9 - 10 | 1294 | 81 | |
NM_175859.2 | NP_787055.1 | 9 - 10 | 1380 | 81 | |
XM_005274562.3 | XP_005274619.1 | 9 - 10 | 1157 | 81 | |
XM_006724503.3 | XP_006724566.1 | 10 - 11 | 1332 | 81 | |
XM_011545545.2 | XP_011543847.1 | 9 - 10 | 1153 | 81 | |
GenBank mRNA | AF226667.1 | - | 9 - 10 | 1197 | 81 |
AK023549.1 | - | 9 - 10 | 1252 | 81 | |
AK024070.1 | - | 9 - 10 | 1235 | 81 | |
AK125332.1 | - | 9 - 10 | 1741 | 81 | |
AK125348.1 | - | 9 - 10 | 1371 | 81 | |
BC006256.2 | - | 7 - 8 | 736 | 81 | |
BC034986.1 | - | 9 - 10 | 1359 | 81 |