The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021930.4 | NP_068749.3 | 8 - 9 | 1343 | 144 |
XM_005250524.3 | XP_005250581.1 | 4 - 5 | 525 | 144 | |
XM_011516458.2 | XP_011514760.1 | 4 - 5 | 498 | 144 | |
XM_017012496.1 | XP_016867985.1 | 8 - 9 | 1122 | 144 | |
XM_017012497.1 | XP_016867986.1 | 8 - 9 | 1214 | 144 | |
XM_017012498.1 | XP_016867987.1 | 5 - 6 | 737 | 144 | |
GenBank mRNA | AF317622.1 | - | 8 - 9 | 1222 | 144 |
AK055508.1 | - | 8 - 9 | 1258 | 144 | |
AK057757.1 | - | 5 - 6 | 744 | 144 | |
AL832825.1 | - | 2 - 3 | 171 | 144 | |
BC068483.1 | - | 8 - 9 | 1344 | 144 |