Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C1orf54
Entrez Gene ID
79630
Gene Name
chromosome 1 open reading frame 54
Gene Aliases
-
Chromosome Location
Chr.1: 150272284 - 150280927 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301039.1 NP_001287968.1 1 - 2 125 100
NM_001301040.1 NP_001287969.1 1 - 2 125 100
XM_011509984.2 XP_011508286.1 1 - 2 125 100
GenBank mRNA T82829.1 - 1 - 2 105 100

View Full Product Details