Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRIQ1
Entrez Gene ID
84125
Gene Name
leucine rich repeats and IQ motif containing 1
Gene Aliases
-
Chromosome Location
Chr.12: 85036321 - 85273004 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079910.1 NP_001073379.1 11 - 12 3000 126
XM_011538817.2 XP_011537119.1 11 - 12 2943 126
XM_011538818.2 XP_011537120.1 11 - 12 2943 126
XM_011538819.2 XP_011537121.1 11 - 12 2943 126
XM_011538823.2 XP_011537125.1 11 - 12 2943 126
XM_011538824.2 XP_011537126.1 11 - 12 2943 126
XM_011538825.2 XP_011537127.1 11 - 12 2943 126
XM_017020019.1 XP_016875508.1 11 - 12 2943 126
GenBank mRNA AB058704.1 - 7 - 8 2564 126
AK307799.1 - 11 - 12 2951 126

View Full Product Details