Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FPR2
Entrez Gene ID
2358
Gene Name
formyl peptide receptor 2
Gene Aliases
ALXR, FMLP-R-II, FMLPX, FPR2A, FPRH1, FPRH2, FPRL1, HM63, LXA4R
Chromosome Location
Chr.19: 51752026 - 51770526 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017026542.1 XP_016882031.1 3 - 4 702 74
GenBank mRNA BC071722.1 - 1 - 2 297 74

View Full Product Details