The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301039.1 | NP_001287968.1 | 6 - 7 | 552 | 86 |
NM_001301040.1 | NP_001287969.1 | 6 - 7 | 549 | 86 | |
NM_001301041.1 | NP_001287970.1 | 4 - 5 | 338 | 86 | |
NM_024579.3 | NP_078855.2 | 5 - 6 | 438 | 86 | |
XM_011509984.2 | XP_011508286.1 | 5 - 6 | 468 | 86 | |
GenBank mRNA | AI004135.1 | - | 1 - 2 | 86 | |
AX770997.1 | - | 5 - 6 | 436 | 86 | |
AX770998.1 | - | 6 - 7 | 466 | 86 | |
BC017761.1 | - | 5 - 6 | 423 | 86 | |
W93038.1 | - | 1 - 2 | 86 |