Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLX4
Entrez Gene ID
84464
Gene Name
SLX4 structure-specific endonuclease subunit
Gene Aliases
BTBD12, FANCP, MUS312
Chromosome Location
Chr.16: 3581181 - 3611598 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032444.2 NP_115820.2 5 - 6 1787 67
XM_011522715.2 XP_011521017.1 5 - 6 1801 67
GenBank mRNA AB075867.1 - 3 - 4 2381 67
BC029952.1 - 4 - 5 1231 67

View Full Product Details