Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC32
Entrez Gene ID
2615
Gene Name
leucine rich repeat containing 32
Gene Aliases
D11S833E, GARP
Chromosome Location
Chr.11: 76657441 - 76670941 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128922.1 NP_001122394.1 1 - 2 129 85
XM_005273902.3 XP_005273959.1 1 - 2 323 85
GenBank mRNA BC052210.1 - 1 - 2 126 85
BC070079.1 - 1 - 2 129 85

View Full Product Details