Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CENPJ
Entrez Gene ID
55835
Gene Name
centromere protein J
Gene Aliases
BM032, CENP-J, CPAP, LAP, LIP1, MCPH6, SASS4, SCKL4, Sas-4
Chromosome Location
Chr.13: 24882274 - 24922889 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018451.4 NP_060921.3 7 - 8 2893 138
NR_047594.1 7 - 8 2893 138
NR_047595.1 7 - 8 2893 138
XM_011535149.2 XP_011533451.1 7 - 8 2893 138
XM_011535150.2 XP_011533452.1 7 - 8 2893 138
XM_017020673.1 XP_016876162.1 7 - 8 2893 138
GenBank mRNA AF139625.1 - 7 - 8 2908 138
AL833182.1 - 7 - 8 2840 138
BC113110.1 - 7 - 8 2829 138
BC113111.1 - 7 - 8 2830 138
BC113662.1 - 7 - 8 2780 138
BC113664.1 - 7 - 8 2780 138
BC143757.1 - 7 - 8 2780 138
BC143758.1 - 7 - 8 2780 138

View Full Product Details