Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR17
Entrez Gene ID
116966
Gene Name
WD repeat domain 17
Gene Aliases
-
Chromosome Location
Chr.4: 176065834 - 176182828 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_170710.4 NP_733828.2 13 - 14 2067 100
NM_181265.3 NP_851782.3 12 - 13 1944 100
XM_005262725.3 XP_005262782.1 11 - 12 1590 100
XM_011531582.2 XP_011529884.1 12 - 13 1932 100
XM_017007686.1 XP_016863175.1 12 - 13 1920 100
XM_017007687.1 XP_016863176.1 13 - 14 2005 100
XM_017007688.1 XP_016863177.1 13 - 14 1922 100
XM_017007689.1 XP_016863178.1 12 - 13 1919 100
XM_017007690.1 XP_016863179.1 13 - 14 2005 100
XM_017007697.1 XP_016863186.1 12 - 13 1921 100
GenBank mRNA AF492460.1 - 13 - 14 1920 100

View Full Product Details