The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_170710.4 | NP_733828.2 | 20 - 21 | 3033 | 123 |
NM_181265.3 | NP_851782.3 | 19 - 20 | 2910 | 123 | |
XM_005262725.3 | XP_005262782.1 | 18 - 19 | 2556 | 123 | |
XM_011531582.2 | XP_011529884.1 | 19 - 20 | 2898 | 123 | |
XM_017007686.1 | XP_016863175.1 | 19 - 20 | 2886 | 123 | |
XM_017007687.1 | XP_016863176.1 | 20 - 21 | 2971 | 123 | |
XM_017007688.1 | XP_016863177.1 | 20 - 21 | 2888 | 123 | |
XM_017007689.1 | XP_016863178.1 | 19 - 20 | 2885 | 123 | |
XM_017007690.1 | XP_016863179.1 | 20 - 21 | 2971 | 123 | |
XM_017007691.1 | XP_016863180.1 | 18 - 19 | 2720 | 123 | |
XM_017007692.1 | XP_016863181.1 | 19 - 20 | 2806 | 123 | |
XM_017007693.1 | XP_016863182.1 | 18 - 19 | 2720 | 123 | |
XM_017007694.1 | XP_016863183.1 | 19 - 20 | 2704 | 123 | |
XM_017007695.1 | XP_016863184.1 | 18 - 19 | 2734 | 123 | |
XM_017007696.1 | XP_016863185.1 | 18 - 19 | 2734 | 123 | |
XM_017007697.1 | XP_016863186.1 | 19 - 20 | 2887 | 123 | |
GenBank mRNA | AF492460.1 | - | 20 - 21 | 2886 | 123 |
AK130128.1 | - | 5 - 6 | 540 | 123 | |
BX648508.1 | - | 4 - 5 | 457 | 123 |