Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR17
Entrez Gene ID
116966
Gene Name
WD repeat domain 17
Gene Aliases
-
Chromosome Location
Chr.4: 176065834 - 176182828 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_170710.4 NP_733828.2 4 - 5 685 122
NM_181265.3 NP_851782.3 3 - 4 562 122
XM_011531582.2 XP_011529884.1 3 - 4 550 122
XM_017007686.1 XP_016863175.1 3 - 4 538 122
XM_017007687.1 XP_016863176.1 3 - 4 551 122
XM_017007688.1 XP_016863177.1 4 - 5 540 122
XM_017007689.1 XP_016863178.1 3 - 4 537 122
XM_017007690.1 XP_016863179.1 3 - 4 551 122
XM_017007691.1 XP_016863180.1 3 - 4 537 122
XM_017007692.1 XP_016863181.1 3 - 4 551 122
XM_017007693.1 XP_016863182.1 3 - 4 537 122
XM_017007694.1 XP_016863183.1 4 - 5 521 122
XM_017007695.1 XP_016863184.1 3 - 4 551 122
XM_017007696.1 XP_016863185.1 3 - 4 551 122
XM_017007697.1 XP_016863186.1 3 - 4 539 122
GenBank mRNA AF492460.1 - 4 - 5 538 122

View Full Product Details