Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TBK1
Entrez Gene ID
29110
Gene Name
TANK binding kinase 1
Gene Aliases
FTDALS4, NAK, T2K
Chromosome Location
Chr.12: 64452060 - 64502119 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013254.3 NP_037386.1 15 - 16 1873 138
XM_005268809.1 XP_005268866.1 15 - 16 1951 138
XM_005268810.1 XP_005268867.1 15 - 16 1933 138
GenBank mRNA AF174536.1 - 14 - 15 1714 138
AF191838.1 - 15 - 16 1776 138
AK002192.1 - 7 - 8 699 138
AK291039.1 - 15 - 16 1818 138
AK303683.1 - 10 - 11 1090 138
BC034950.2 - 15 - 16 138

View Full Product Details