The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005232.4 | NP_005223.4 | 3 - 4 | 521 | 69 |
XM_006715880.3 | XP_006715943.1 | 3 - 4 | 494 | 69 | |
GenBank mRNA | AK290351.1 | - | 3 - 4 | 485 | 69 |
AU131228.1 | - | 3 - 4 | 480 | 69 | |
BC130291.1 | - | 3 - 4 | 505 | 69 | |
BC144156.1 | - | 3 - 4 | 505 | 69 | |
CN289899.1 | - | 1 - 2 | 198 | 69 | |
EU826604.1 | - | 3 - 4 | 434 | 69 | |
EU826605.1 | - | 3 - 4 | 434 | 69 | |
M18391.1 | - | 3 - 4 | 527 | 69 |