The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277145.1 | NP_001264074.1 | 1 - 2 | 262 | 68 |
GenBank mRNA | AK056913.1 | - | 1 - 2 | 262 | 68 |